Assay Details
Gene Symbol: DYX1C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008674.1 | NP_001008674.1 | 2 - 3 | 361 | 132 | |
GenBank mRNA | AY464141.1 | - | 2 - 3 | 361 | 132 | |
CR389904.1 | - | 2 - 3 | 339 | 132 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
