Assay Details
Gene Symbol: DYX1C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008674.1 | NP_001008674.1 | 8 - 9 | 1254 | 76 | |
GenBank mRNA | AY464141.1 | - | 8 - 9 | 1254 | 76 | |
CR387522.1 | - | 3 - 4 | 363 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
