Assay Details
| Gene Symbol: CHAF1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008677.2 | NP_001008677.2 | 6 - 7 | 696 | 65 | |
| XM_015299470.2 | XP_015154956.1 | 6 - 7 | 628 | 65 | ||
| XM_015299476.2 | XP_015154962.1 | 6 - 7 | 3041 | 65 | ||
| XM_025149509.1 | XP_025005277.1 | 6 - 7 | 670 | 65 | ||
| GenBank mRNA | AB195693.1 | - | 5 - 6 | 581 | 65 | |
| AJ719705.1 | - | 6 - 7 | 696 | 65 | ||
More Information
Set Membership: |
|

CHAF1B
Genomic Map