Assay Details
Gene Symbol: PLAU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205443.2 | NP_990774.2 | 5 - 6 | 478 | 57 | |
GenBank mRNA | J05187.1 | - | 5 - 6 | 478 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
