Assay Details
| Gene Symbol: TNNC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205450.2 | NP_990781.1 | 4 - 5 | 366 | 59 | |
| GenBank mRNA | M19027.1 | - | 4 - 5 | 370 | 59 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TNNC2| Gene Symbol: TNNC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205450.2 | NP_990781.1 | 4 - 5 | 366 | 59 | |
| GenBank mRNA | M19027.1 | - | 4 - 5 | 370 | 59 | |
Set Membership: |
|