Assay Details
Gene Symbol: MLNR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127313.1 | NP_001120785.1 | 1 - 2 | 797 | 142 | |
GenBank mRNA | EU122384.1 | - | 1 - 2 | 797 | 142 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
