Assay Details
| Gene Symbol: PLCXD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128637.1 | NP_001122109.1 | 3 - 4 | 303 | 79 | |
| XM_015277705.2 | XP_015133191.1 | 3 - 4 | 347 | 79 | ||
| XM_015277706.2 | XP_015133192.1 | 3 - 4 | 373 | 79 | ||
| GenBank mRNA | AJ720680.1 | - | 3 - 4 | 352 | 79 | |
More Information
Set Membership: |
|

PLCXD1
Genomic Map