Assay Details
| Gene Symbol: PNPLA8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130737.1 | NP_001124209.1 | 8 - 9 | 2131 | 62 | |
| XM_015281155.1 | XP_015136641.1 | 9 - 10 | 2431 | 62 | ||
| XM_015281161.2 | XP_015136647.1 | 9 - 10 | 2341 | 62 | ||
| XM_015281171.2 | XP_015136657.1 | 9 - 10 | 2367 | 62 | ||
| XM_025147679.1 | XP_025003447.1 | 9 - 10 | 2239 | 62 | ||
| GenBank mRNA | EU419878.1 | - | 8 - 9 | 2131 | 62 | |
More Information
Set Membership: |
|

PNPLA8
Genomic Map