Assay Details
| Gene Symbol: DMB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135166.1 | NP_001128638.1 | 4 - 5 | 731 | 78 | |
| XM_015294966.2 | XP_015150452.1 | 3 - 4 | 901 | 78 | ||
| XM_015294967.2 | XP_015150453.1 | 4 - 5 | 1089 | 78 | ||
| GenBank mRNA | BX932520.2 | - | 4 - 5 | 734 | 78 | |
| CR405752.1 | - | 4 - 5 | 715 | 78 | ||
More Information
Set Membership: |
|

DMB2
Genomic Map