Assay Details
| Gene Symbol: GSPT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135677.1 | NP_001129149.1 | 1 - 2 | 430 | 147 | |
| GenBank mRNA | AJ719783.1 | - | 1 - 2 | 398 | 147 | |
More Information
Set Membership: |
|

GSPT2
Genomic Map