Assay Details
Gene Symbol: GSPT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135677.1 | NP_001129149.1 | 2 - 3 | 488 | 86 | |
XM_015294247.2 | XP_015149733.1 | 2 - 3 | 190 | 86 | ||
GenBank mRNA | AJ719783.1 | - | 2 - 3 | 456 | 86 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015294247.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
