Assay Details
| Gene Symbol: GSPT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135677.1 | NP_001129149.1 | 3 - 4 | 523 | 123 | |
| XM_015294247.2 | XP_015149733.1 | 3 - 4 | 225 | 123 | ||
| GenBank mRNA | AJ719783.1 | - | 3 - 4 | 491 | 123 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015294247.2 transcripts.
Set Membership: |
|

GSPT2
Genomic Map