Assay Details
| Gene Symbol: GSPT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135677.1 | NP_001129149.1 | 5 - 6 | 791 | 123 | |
| XM_015294247.2 | XP_015149733.1 | 5 - 6 | 493 | 123 | ||
More Information
Set Membership: |
|

GSPT2
Genomic Map