Assay Details
| Gene Symbol: PROKR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145229.1 | NP_001138701.1 | 2 - 3 | 761 | 90 | |
| XM_015278042.2 | XP_015133528.1 | 2 - 3 | 867 | 90 | ||
| GenBank mRNA | FJ619534.1 | - | 2 - 3 | 761 | 90 | |
More Information
Set Membership: |
|

PROKR1
Genomic Map