Assay Details
Gene Symbol: POP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145914.1 | NP_001139386.1 | 4 - 5 | 388 | 79 | |
XM_025154219.1 | XP_025009987.1 | 4 - 5 | 1210 | 79 | ||
XM_025154220.1 | XP_025009988.1 | 4 - 5 | 1210 | 79 | ||
XM_025154221.1 | XP_025009989.1 | 4 - 5 | 481 | 79 | ||
GenBank mRNA | BU278500.1 | - | 2 - 3 | 171 | 79 | |
BX930910.2 | - | 3 - 4 | 364 | 79 | ||
BX931329.2 | - | 4 - 5 | 383 | 79 | ||
BX932536.1 | - | 1 - 2 | 417 | 79 | ||
BX934072.2 | - | 3 - 4 | 409 | 79 | ||
BX934249.1 | - | 3 - 4 | 418 | 79 | ||
CR405732.1 | - | 4 - 5 | 388 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
