Assay Details
| Gene Symbol: H3F3C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031482.2 | NP_001026653.2 | 2 - 3 | 309 | 57 | |
| XM_015283737.2 | XP_015139223.2 | 3 - 4 | 351 | 57 | ||
| GenBank mRNA | AJ441927.1 | - | 3 - 4 | 389 | 57 | |
| AM071891.1 | - | 1 - 2 | 69 | 57 | ||
More Information
Set Membership: |
|

H3F3C
Genomic Map