Assay Details
| Gene Symbol: FEN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079491.1 | NP_001072959.1 | 1 - 2 | 33 | 57 | |
| GenBank mRNA | AJ719700.1 | - | 1 - 2 | 33 | 57 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AJ719700.1, NM_001079491.1 transcripts.
Set Membership: |
|

FEN1
Genomic Map