Assay Details
| Gene Symbol: VAMP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031121.1 | NP_001026292.1 | 9 | 2747 | 93 | |
| XM_015278285.2 | XP_015133771.1 | 9 | 2598 | 93 | ||
| XM_015278286.2 | XP_015133772.1 | 9 | 2660 | 93 | ||
| XM_015278287.2 | XP_015133773.1 | 9 | 2619 | 93 | ||
| XM_015278288.2 | XP_015133774.1 | 9 | 2580 | 93 | ||
| XM_015278289.2 | XP_015133775.1 | 9 | 2591 | 93 | ||
| GenBank mRNA | AJ719860.1 | - | 9 | 2747 | 93 | |
More Information
Set Membership: |
|

VAMP7
Genomic Map