Assay Details
Gene Symbol: CDCA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031212.2 | NP_001026383.2 | 3 | 1352 | 80 | |
XM_015287486.2 | XP_015142972.1 | 4 | 1650 | 80 | ||
XM_015287487.2 | XP_015142973.1 | 3 | 1314 | 80 | ||
XM_015287489.2 | XP_015142975.1 | 3 | 1341 | 80 | ||
XM_025150796.1 | XP_025006564.1 | 3 | 1380 | 80 | ||
XM_025150797.1 | XP_025006565.1 | 3 | 3870 | 80 | ||
GenBank mRNA | AJ719887.1 | - | 3 | 1352 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
