Assay Details
| Gene Symbol: TSC22D4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077234.1 | NP_001070702.1 | 3 | 749 | 108 | |
| XM_015278317.2 | XP_015133803.1 | 3 | 1022 | 108 | ||
| XM_015278318.2 | XP_015133804.1 | 3 | 760 | 108 | ||
| GenBank mRNA | BX929628.1 | - | 3 | 783 | 108 | |
| BX932583.1 | - | 4 | 692 | 108 | ||
| BX936258.1 | - | 3 | 775 | 108 | ||
| CR406406.1 | - | 1 | 454 | 108 | ||
| DQ917420.1 | - | 3 | 749 | 108 | ||
More Information
Set Membership: |
|

TSC22D4
Genomic Map