Assay Details
| Gene Symbol: RGS19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204229.1 | NP_989560.1 | 6 | 1952 | 99 | |
| XM_025142157.1 | XP_024997925.1 | 4 | 1715 | 99 | ||
| GenBank mRNA | AF502147.1 | - | 6 | 1952 | 99 | |
More Information
Set Membership: |
|

RGS19
Genomic Map