Assay Details
| Gene Symbol: NUDT16L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204780.1 | NP_990111.1 | 3 | 1052 | 101 | |
| GenBank mRNA | AF095446.1 | - | 3 | 1052 | 101 | |
| CR353581.1 | - | 3 | 1057 | 101 | ||
| CR386149.1 | - | 2 | 684 | 101 | ||
More Information
Set Membership: |
|

NUDT16L1
Genomic Map