Assay Details
Gene Symbol: SPRY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204800.1 | NP_990131.1 | 2 | 553 | 73 | |
XM_015275115.2 | XP_015130601.1 | 2 | 583 | 73 | ||
XM_015275119.2 | XP_015130605.1 | 2 | 593 | 73 | ||
XM_015275123.2 | XP_015130609.1 | 2 | 543 | 73 | ||
GenBank mRNA | AF176904.1 | - | 2 | 553 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
