Assay Details
| Gene Symbol: P2RY1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205333.1 | NP_990664.1 | 2 | 1251 | 86 | |
| GenBank mRNA | X73268.1 | - | 1251 | 86 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

P2RY1| Gene Symbol: P2RY1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205333.1 | NP_990664.1 | 2 | 1251 | 86 | |
| GenBank mRNA | X73268.1 | - | 1251 | 86 | ||
Set Membership: |
|