Assay Details
Gene Symbol: SST | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205336.1 | NP_990667.1 | 2 | 454 | 86 | |
GenBank mRNA | EF593949.1 | - | 2 | 432 | 86 | |
X60191.1 | - | 471 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
