Assay Details
Gene Symbol: ACHE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_025146106.1 | XP_025001874.1 | 6 | 2387 | 103 | |
GenBank mRNA | U03472.1 | - | 2345 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
