Assay Details
| Gene Symbol: OPN1LW | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205440.1 | NP_990771.1 | 1153 | 91 | ||
| GenBank mRNA | M62903.1 | - | 1153 | 91 | ||
| X57490.1 | - | 1152 | 91 | |||
More Information
Set Membership: |
|

OPN1LW
Genomic Map