Assay Details
| Gene Symbol: LOC771456 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001318432.1 | NP_001305361.1 | 6 - 7 | 524 | 105 | |
| XM_004943000.3 | XP_004943057.1 | 5 - 6 | 1542 | 105 | ||
| XM_025152266.1 | XP_025008034.1 | 7 - 8 | 776 | 105 | ||
| XM_025152267.1 | XP_025008035.1 | 7 - 8 | 904 | 105 | ||
| GenBank mRNA | KF271750.1 | - | 6 - 7 | 524 | 105 | |
More Information
Set Membership: |
|

LOC771456
Genomic Map