Assay Details
Gene Symbol: RSPO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001318444.1 | NP_001305373.1 | 2 - 3 | 285 | 86 | |
XM_004947756.3 | XP_004947813.1 | 5 - 6 | 856 | 86 | ||
XM_004947757.3 | XP_004947814.1 | 3 - 4 | 1002 | 86 | ||
XM_025142990.1 | XP_024998758.1 | 3 - 4 | 1066 | 86 | ||
XM_417760.6 | XP_417760.3 | 3 - 4 | 734 | 86 | ||
GenBank mRNA | KC858867.1 | - | 2 - 3 | 285 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
