Assay Details
Gene Symbol: TMEM8C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001318457.1 | NP_001305386.1 | 2 - 3 | 309 | 114 | |
GenBank mRNA | BX931401.1 | - | 2 - 3 | 286 | 114 | |
BX934605.2 | - | 2 - 3 | 295 | 114 | ||
KP230536.1 | - | 2 - 3 | 309 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
