Assay Details
Gene Symbol: WBP2NL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001318465.1 | NP_001305394.1 | 1 - 2 | 79 | 59 | |
XM_001233671.5 | XP_001233672.1 | 1 - 2 | 92 | 59 | ||
XM_015289101.2 | XP_015144587.1 | 1 - 2 | 92 | 59 | ||
GenBank mRNA | AJ851511.1 | - | 1 - 2 | 79 | 59 | |
CR386295.1 | - | 1 - 2 | 76 | 59 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015289101.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
