Assay Details
| Gene Symbol: IL2RB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001318989.1 | NP_001305918.1 | 6 - 7 | 647 | 63 | |
| GenBank mRNA | JN642526.1 | - | 6 - 7 | 647 | 63 | |
More Information
Set Membership: |
|

IL2RB
Genomic Map