Assay Details
| Gene Symbol: ZAR1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001171543.2 | NP_001165014.2 | 1 - 2 | 1176 | 75 | |
| GenBank mRNA | FJ914588.1 | - | 1 - 2 | 587 | 75 | |
More Information
Set Membership: |
|

ZAR1L
Genomic Map