Assay Details
Gene Symbol: LARP6L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001326492.1 | NP_001313421.1 | 1 - 2 | 106 | 95 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001326492.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
