Assay Details
| Gene Symbol: CYP2AB4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001329496.1 | NP_001316425.1 | 8 - 9 | 1654 | 89 | |
| XM_426708.6 | XP_426708.3 | 9 - 10 | 1403 | 89 | ||
More Information
Set Membership: |
|

CYP2AB4
Genomic Map