Assay Details
| Gene Symbol: CYP2AC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001329498.1 | NP_001316427.1 | 2 - 3 | 491 | 119 | |
| XM_025148803.1 | XP_025004571.1 | 2 - 3 | 487 | 119 | ||
More Information
Set Membership: |
|

CYP2AC1
Genomic Map