Assay Details
Gene Symbol: TPM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001245927.2 | NP_001232856.1 | 6 - 7 | 664 | 81 | |
NM_001352039.1 | NP_001338968.1 | 6 - 7 | 664 | 81 | ||
NM_001352040.1 | NP_001338969.1 | 6 - 7 | 664 | 81 | ||
NM_001352041.1 | NP_001338970.1 | 7 - 8 | 751 | 81 | ||
NM_001352042.1 | NP_001338971.1 | 6 - 7 | 664 | 81 | ||
NM_001352043.1 | NP_001338972.1 | 6 - 7 | 664 | 81 | ||
XM_015298565.1 | XP_015154051.1 | 7 - 8 | 754 | 81 | ||
XM_015298568.1 | XP_015154054.1 | 6 - 7 | 661 | 81 | ||
XM_015298569.2 | XP_015154055.1 | 6 - 7 | 661 | 81 | ||
XM_015298570.1 | XP_015154056.1 | 7 - 8 | 754 | 81 | ||
XM_015298571.1 | XP_015154057.1 | 7 - 8 | 754 | 81 | ||
XM_015298576.1 | XP_015154062.1 | 7 - 8 | 722 | 81 | ||
XM_015298577.1 | XP_015154063.1 | 7 - 8 | 722 | 81 | ||
XM_015298578.1 | XP_015154064.1 | 7 - 8 | 754 | 81 | ||
XM_015298582.1 | XP_015154068.1 | 6 - 7 | 661 | 81 | ||
XM_015298585.1 | XP_015154071.1 | 6 - 7 | 661 | 81 | ||
GenBank mRNA | AJ719737.1 | - | 6 - 7 | 666 | 81 | |
CN232609.1 | - | 6 - 7 | 621 | 81 | ||
CR389053.1 | - | 1 - 2 | 555 | 81 | ||
DR424358.1 | - | 6 - 7 | 600 | 81 | ||
MF347378.1 | - | 7 - 8 | 720 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
