Assay Details
| Gene Symbol: PACSIN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159511.1 | NP_001152983.1 | 5 - 6 | 715 | 68 | |
| NM_205089.2 | NP_990420.2 | 5 - 6 | 715 | 68 | ||
| XM_015290048.2 | XP_015145534.1 | 5 - 6 | 745 | 68 | ||
| XM_015290052.2 | XP_015145538.1 | 5 - 6 | 723 | 68 | ||
| XM_015290053.2 | XP_015145539.1 | 5 - 6 | 617 | 68 | ||
| XM_015290057.2 | XP_015145543.1 | 5 - 6 | 718 | 68 | ||
| XM_015290064.2 | XP_015145550.1 | 5 - 6 | 617 | 68 | ||
| XM_015290070.2 | XP_015145556.1 | 5 - 6 | 707 | 68 | ||
| XM_015290076.2 | XP_015145562.1 | 5 - 6 | 707 | 68 | ||
| XM_015290083.2 | XP_015145569.1 | 5 - 6 | 707 | 68 | ||
| XM_015290091.2 | XP_015145577.1 | 4 - 5 | 634 | 68 | ||
| XM_025146493.1 | XP_025002261.1 | 5 - 6 | 725 | 68 | ||
| GenBank mRNA | CR385642.1 | - | 5 - 6 | 712 | 68 | |
| Z50798.1 | - | 681 | 68 | |||
More Information
Set Membership: |
|

PACSIN2
Genomic Map