Assay Details
| Gene Symbol: E2F1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205219.1 | NP_990550.1 | 3 - 4 | 634 | 101 | |
| XM_015296338.2 | XP_015151824.1 | 3 - 4 | 605 | 101 | ||
| GenBank mRNA | X89245.1 | - | 634 | 101 | ||
More Information
Set Membership: |
|

E2F1
Genomic Map