Assay Details
| Gene Symbol: BZFP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205309.3 | NP_990640.3 | 2 - 3 | 230 | 65 | |
| XM_015294946.2 | XP_015150432.2 | 2 - 3 | 253 | 65 | ||
| GenBank mRNA | X15538.1 | - | 210 | 65 | ||
| Gene Symbol: LOC112533547 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025155836.1 | XP_025011604.1 | 2 - 3 | 177 | 65 | |
More Information
Set Membership: |
|

BZFP1
Genomic Map