Assay Details
| Gene Symbol: FAM46C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007972.1 | NP_001007973.1 | 1 - 2 | 65 | 103 | |
| GenBank mRNA | AJ719839.1 | - | 1 - 2 | 65 | 103 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AJ719839.1, NM_001007972.1 transcripts.
Set Membership: |
|

FAM46C
Genomic Map