Assay Details
| Gene Symbol: CCL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030360.2 | NP_001025531.2 | 1 - 2 | 132 | 84 | |
| GenBank mRNA | AJ243034.1 | - | 1 - 2 | 132 | 84 | |
| CR354001.1 | - | 1 - 2 | 130 | 84 | ||
| CR389044.1 | - | 1 - 2 | 126 | 84 | ||
More Information
Set Membership: |
|

CCL4
Genomic Map