Assay Details
| Gene Symbol: MOB4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031557.2 | NP_001026728.2 | 3 - 4 | 248 | 72 | |
| GenBank mRNA | AJ851405.1 | - | 3 - 4 | 230 | 72 | |
More Information
Set Membership: |
|

MOB4
Genomic Map