Assay Details
| Gene Symbol: CCR7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198752.1 | NP_001185681.1 | 1 - 2 | 78 | 79 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001198752.1 transcripts.
Set Membership: |
|

CCR7
Genomic Map