Assay Details
| Gene Symbol: RDH10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199459.1 | NP_001186388.1 | 5 - 6 | 1135 | 65 | |
| XM_015282780.2 | XP_015138266.1 | 5 - 6 | 1184 | 65 | ||
| GenBank mRNA | CR382432.1 | - | 5 - 6 | 1165 | 65 | |
More Information
Set Membership: |
|

RDH10
Genomic Map