Assay Details
Gene Symbol: CCSER2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199508.1 | NP_001186437.1 | 3 - 4 | 1733 | 86 | |
XM_015287966.2 | XP_015143452.1 | 5 - 6 | 2183 | 86 | ||
XM_015287968.2 | XP_015143454.1 | 5 - 6 | 2080 | 86 | ||
XM_015287969.2 | XP_015143455.1 | 4 - 5 | 1904 | 86 | ||
XM_015287971.2 | XP_015143457.1 | 5 - 6 | 2183 | 86 | ||
XM_015287972.2 | XP_015143458.1 | 5 - 6 | 2183 | 86 | ||
XM_015287973.2 | XP_015143459.1 | 5 - 6 | 2183 | 86 | ||
XM_015287974.2 | XP_015143460.1 | 3 - 4 | 526 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
