Assay Details
| Gene Symbol: SIGIRR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199542.1 | NP_001186471.1 | 3 - 4 | 392 | 54 | |
| XM_015286201.2 | XP_015141687.1 | 3 - 4 | 442 | 54 | ||
| XM_015286202.2 | XP_015141688.1 | 3 - 4 | 430 | 54 | ||
| XM_025150652.1 | XP_025006420.1 | 3 - 4 | 486 | 54 | ||
| XM_025150653.1 | XP_025006421.1 | 3 - 4 | 473 | 54 | ||
| XM_025150654.1 | XP_025006422.1 | 2 - 3 | 1675 | 54 | ||
| XM_025150655.1 | XP_025006423.1 | 3 - 4 | 485 | 54 | ||
More Information
Set Membership: |
|

SIGIRR
Genomic Map