Assay Details
| Gene Symbol: RNLS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199599.1 | NP_001186528.1 | 4 - 5 | 507 | 75 | |
| XM_015278676.2 | XP_015134162.1 | 4 - 5 | 610 | 75 | ||
| XM_015278677.2 | XP_015134163.1 | 3 - 4 | 608 | 75 | ||
| XM_015278678.2 | XP_015134164.1 | 3 - 4 | 540 | 75 | ||
| XM_015278679.2 | XP_015134165.1 | 4 - 5 | 2411 | 75 | ||
| GenBank mRNA | BX935542.2 | - | 4 - 5 | 504 | 75 | |
More Information
Set Membership: |
|

RNLS
Genomic Map