Assay Details
Gene Symbol: STON1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204865.2 | NP_001191794.2 | 2 - 3 | 2124 | 75 | |
XM_015278009.2 | XP_015133495.1 | 3 - 4 | 2222 | 75 | ||
XM_015278010.2 | XP_015133496.1 | 4 - 5 | 2398 | 75 | ||
XM_015278011.2 | XP_015133497.1 | 3 - 4 | 2313 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
