Assay Details
| Gene Symbol: COX19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001256807.1 | NP_001243736.1 | 1 - 2 | 122 | 88 | |
| GenBank mRNA | BU382349.1 | - | 1 - 2 | 95 | 88 | |
More Information
Set Membership: |
|

COX19
Genomic Map